(3S)-(+)-1-Benzyl-3-aminopyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Benzyl-3-aminopyrrolidine, (S)-(+)-1-Benzyl-3-aminopyrrolidine

A207644-100G A207644-1G A207644-5G
76770-362EA 470 USD
76770-362 76770-364 76770-366
(3S)-(+)-1-Benzyl-3-aminopyrrolidine 97%
(3S)-(+)-1-Benzyl-3-aminopyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂
MDL Number: MFCD00082766
CAS Number: 114715-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR