(2,4-Difluorophenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A206106-100G A206106-10G A206106-25G A206106-5G
76770-340EA 62.99 USD
76770-340 76770-342 76770-344 76770-346
(2,4-Difluorophenyl)acetonitrile 97%
(2,4-Difluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MDL Number: MFCD00009971
CAS Number: 656-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR