4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(((Benzyloxy)carbonyl)amino)-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid

A206061-1G A206061-250MG A206061-5G
76770-334EA 50.93 USD
76770-334 76770-336 76770-338
4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid 97%
4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₂₆N₂O₆
MW: 378.42 g/mol
MDL Number: MFCD02683056
CAS Number: 288154-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR