6-Bromo-5-iodopyridin-3-amine 97%
Supplier: Ambeed
|
Warning
|
A206050-1G
A206050-250MG
A206050-25G
A206050-5G
76770-326EA
15
USD
76770-326
76770-328
76770-330
76770-332
6-Bromo-5-iodopyridin-3-amine 97%
6-Bromo-5-iodopyridin-3-amine
Precaución: for Research Use Only
Formula:
C₅H₄BrIN₂ |
MDL Number:
MFCD09038022 CAS Number: 697300-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...