5-Dimethylaminothiophene-2-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Dimethylamino)thiophene-2-carbaldehyde

A205864-100MG A205864-1G A205864-250MG A205864-5G
76770-316EA 38.2 USD
76770-316 76770-318 76770-320 76770-322
5-Dimethylaminothiophene-2-carbaldehyde 98%
5-Dimethylaminothiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₉NOS
MW: 155.22 g/mol
MDL Number: MFCD00708772
CAS Number: 24372-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR