tert-Butyl 5-bromonicotinate 97%
Supplier: Ambeed
|
Warning
|
A205697-100MG
A205697-1G
A205697-250MG
A205697-5G
76770-306EA
11
USD
76770-306
76770-308
76770-310
76770-312
tert-Butyl 5-bromonicotinate 97%
tert-Butyl 5-bromonicotinate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂BrNO₂ MW: 258.112 g/mol |
MDL Number:
MFCD07367903 CAS Number: 263270-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...