3,4-Diisopropoxy-3-cyclobutene-1,2-dione 98%

Supplier: Ambeed
Warning

Synonyms: Diisopropyl Squarate, 3,4-Diisopropoxycyclobut-3-ene-1,2-dione

A204206-100G A204206-1G A204206-25G A204206-5G
76770-268EA 285.39 USD
76770-268 76770-270 76770-272 76770-274
3,4-Diisopropoxy-3-cyclobutene-1,2-dione 98%
3,4-Diisopropoxy-3-cyclobutene-1,2-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
MDL Number: MFCD00075125
CAS Number: 61699-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR