Sodium 2,3-dimercapto-1-propanesulfonate 98%

Supplier: Ambeed
Danger

A203975-1G A203975-5G
76770-244EA 88.06 USD
76770-244 76770-246
Sodium 2,3-dimercapto-1-propanesulfonate 98%
Sodium 2,3-dimercapto-1-propanesulfonate

Precaución: for Research Use Only

Formula: C₃H₇NaO₃S₃
MW: 210.27 g/mol
MDL Number: MFCD00007523
CAS Number: 4076-02-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR