3-Bromo-5-nitro-1H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-nitroindazole

A203382-100G A203382-10G A203382-500G A203382-5G
76770-230EA 165.51 USD
76770-230 76770-232 76770-234 76770-236
3-Bromo-5-nitro-1H-indazole 97%
3-Bromo-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
MDL Number: MFCD07367964
CAS Number: 67400-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR