3,4'-Diaminodiphenyl ether 97%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Aminophenoxy)aniline, 3,4'-Oxydianiline

A203027-10G A203027-25G A203027-100G A203027-500G
76770-228EA 6 USD
76770-228 77086-198 77567-938 77567-940
3,4'-Diaminodiphenyl ether 97%
3,4'-Diaminodiphenyl ether

Precaución: For research use only.

Formula: C₁₂H₁₂N₂O
MW: 200.2365 g/mol
MDL Number: MFCD00036097
CAS Number: 2657-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR