Ethyl (ethoxymethylene)cyanoacetate 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-Cyano-3-Ethoxyacrylate

A202964-100G A202964-500G A202964-5G A202964-25G
76770-220EA 78.51 USD
76770-220 76770-222 76770-224 76770-226 77302-838
Ethyl (ethoxymethylene)cyanoacetate 98%
Ethyl (ethoxymethylene)cyanoacetate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MDL Number: MFCD00009136
CAS Number: 94-05-3
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR