2-Fluoro-4-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A202955-1G A202955-25G A202955-5G A202955-100G
76770-218EA 6 USD
76770-218 77567-934 77598-300 77877-631
2-Fluoro-4-hydroxybenzoic acid 98%
2-Fluoro-4-hydroxybenzoic acid

Precaución: For research use only.

Formula: C₇H₅FO₃
MDL Number: MFCD01310985
CAS Number: 65145-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR