6-Chloro[1,2,4]triazolo[4,3-b]pyridazine 97%

Supplier: Ambeed
Warning

A201888-1G A201888-250MG A201888-25G A201888-5G
76770-166EA 24 USD
76770-166 76770-168 76770-170 76770-172
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine 97%
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine

Precaución: for Research Use Only

Formula: C₅H₃ClN₄
MDL Number: MFCD00219047
CAS Number: 28593-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR