2-Bromo-5-fluoro-4-methylaniline 98%

Supplier: Ambeed
Warning

A201445-100G A201445-1G A201445-25G A201445-5G
76770-158EA 960 USD
76770-158 76770-160 76770-162 76770-164
2-Bromo-5-fluoro-4-methylaniline 98%
2-Bromo-5-fluoro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD00142871
CAS Number: 202865-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR