5-Phenyl-o-toluidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-[1,1'-biphenyl]-3-amine, 3-Amino-4-methylbiphenyl

A201076-5G A201076-250MG
76770-152EA 206.88 USD
76770-152 77625-998
5-Phenyl-o-toluidine 95%
5-Phenyl-o-toluidine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.26 g/mol
MDL Number: MFCD00270126
CAS Number: 80938-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR