Mexiletine hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: 1-(2,6-Dimethylphenoxy)propan-2-amine hydrochloride, 1-(2,6-Dimethylphenoxy)-2-propylamine Hydrochloride, Mexiletine HCl

A200751-100MG A200751-25MG A200751-25G A200751-5G
76770-140EA 8 USD
76770-140 76770-142 77877-604 77877-605
Mexiletine hydrochloride 99%
Mexiletine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO
MW: 215.72 g/mol
MDL Number: MFCD00216024
CAS Number: 5370-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR