4-Fluoro-m-toluonitrile 98%

Supplier: Ambeed
Warning

A199638-10G A199638-25G
76770-106EA 9.55 USD
76770-106 76770-108
4-Fluoro-m-toluonitrile 98%
4-Fluoro-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03788510
CAS Number: 185147-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR