ω-Chloro-2,5-difluoroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-2',5'-Difluoroacetophenone

A199178-25G A199178-5G
76770-090EA 78.51 USD
76770-090 76770-092
ω-Chloro-2,5-difluoroacetophenone 98%
ω-Chloro-2,5-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O
MW: 190.5745 g/mol
MDL Number: MFCD04115827
CAS Number: 60468-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR