1-Bromo-2,5-bis(trifluoromethyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Bis(trifluoromethyl)bromobenzene, 2-Bromo-1,4-bis(trifluoromethyl)benzene

A199001-10G A199001-1G A199001-25G A199001-5G A199001-100G
76770-082EA 15.92 USD
76770-082 76770-084 76770-086 76770-088 77625-984
1-Bromo-2,5-bis(trifluoromethyl)benzene 98%
1-Bromo-2,5-bis(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₃BrF₆
MW: 293.01 g/mol
MDL Number: MFCD03094336
CAS Number: 7617-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR