5H-Cyclopenta[c]pyridin-7(6H)-one 95%

Supplier: Ambeed
Warning

A198482-100MG A198482-1G A198482-250MG
76770-070EA 95.49 USD
76770-070 76770-072 76770-074
5H-Cyclopenta[c]pyridin-7(6H)-one 95%
5H-Cyclopenta[c]pyridin-7(6H)-one

Precaución: for Research Use Only

Formula: C₈H₇NO
MW: 133.14 g/mol
MDL Number: MFCD08694510
CAS Number: 51907-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR