3-(Hydroxymethyl)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: Biphenyl-3-methanol, [1,1'-Biphenyl]-3-ylmethanol, 3-Phenylbenzyl Alcohol

A198425-1G A198425-250MG A198425-25G A198425-5G
76770-062EA 50.93 USD
76770-062 76770-064 76770-066 76770-068
3-(Hydroxymethyl)biphenyl 98%
3-(Hydroxymethyl)biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
MDL Number: MFCD08703631
CAS Number: 69605-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR