(4-(Morpholinomethyl)phenyl)methanol 98%

Supplier: Ambeed
Warning

A198237-100MG A198237-1G A198237-250MG A198237-5G
76770-048EA 36.08 USD
76770-048 76770-050 76770-052 76770-054
(4-(Morpholinomethyl)phenyl)methanol 98%
(4-(Morpholinomethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.26 g/mol
MDL Number: MFCD04974050
CAS Number: 91271-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR