5-Chloro-2,4-difluorobenzonitrile 98%

Supplier: Ambeed
Warning

A196817-10G A196817-1G A196817-250MG A196817-25G A196817-5G
76769-998EA 175.05 USD
76769-998 76770-000 76770-002 76770-004 76770-006
5-Chloro-2,4-difluorobenzonitrile 98%
5-Chloro-2,4-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₂N
MW: 173.547 g/mol
MDL Number: MFCD06659465
CAS Number: 146780-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR