6-Bromo-2,3-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A196752-10G A196752-1G A196752-25G A196752-5G
76769-990EA 58.35 USD
76769-990 76769-992 76769-994 76769-996
6-Bromo-2,3-difluorobenzaldehyde 98%
6-Bromo-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MDL Number: MFCD07368119
CAS Number: 360576-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR