3,4,5-Trifluorobenzonitrile 98%

Supplier: Ambeed
Danger

A195610-10G A195610-5G
76769-966EA 11.67 USD
76769-966 77625-968
3,4,5-Trifluorobenzonitrile 98%
3,4,5-Trifluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂F₃N
MW: 157.09 g/mol
MDL Number: MFCD00074948
CAS Number: 134227-45-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR