Methyl-N-phenylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: N-Phenylcarbamic acid Methyl Ester, Methyl N-Phenylcarbamate

A194993-25G A194993-5G
76769-962EA 47 USD
76769-962 76769-964
Methyl-N-phenylcarbamate 98%
Methyl-N-phenylcarbamate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD00027552
CAS Number: 2603-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR