3-Cyano-4-methylaniline 97%
Supplier: Ambeed
|
Warning
|
A193033-100G
A193033-10G
A193033-25G
A193033-5G
76769-932EA
107
USD
76769-932
76769-934
76769-936
76769-938
76769-940
3-Cyano-4-methylaniline 97%
3-Cyano-4-methylaniline
Precaución: for Research Use Only
Formula:
C₈H₈N₂ |
MDL Number:
MFCD00017608 CAS Number: 50670-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...