5'-Trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid

A192947-250MG A192947-5G
76769-926EA 8.49 USD
76769-926 77598-254
5'-Trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-carboxylic acid 97%
5'-Trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃N₂O₂
MW: 274.24 g/mol
MDL Number: MFCD00209248
CAS Number: 406476-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR