5H-[1,3]Dioxolo[4,5-f]indole 97%

Supplier: Ambeed
Warning

A192188-100MG A192188-1G A192188-250MG A192188-5G A192188-25G
76769-898EA 9.55 USD
76769-898 76769-900 76769-902 77593-226 77625-920
5H-[1,3]Dioxolo[4,5-f]indole 97%
5H-[1,3]Dioxolo[4,5-f]indole

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.15 g/mol
MDL Number: MFCD00046910
CAS Number: 267-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR