Ferulic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-hydroxy-3-methoxycinnamate, Methyl 3-(4-hydroxy-3-methoxyphenyl)acrylate

A192063-1G A192063-25G A192063-5G A192063-100G A192063-10G
76769-892EA 6.37 USD
76769-892 76769-894 76769-896 77598-244 77598-246
Ferulic acid methyl ester 98%
Ferulic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.22 g/mol
MDL Number: MFCD00017208
CAS Number: 2309-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR