Valrubicin (AD-32) 98%

Supplier: Ambeed
Danger

Synonyms: 2-oxo-2-((2S,4S)-2,5,12-trihydroxy-4-(((2R,4S,5S,6S)-5-hydroxy-6-methyl-4-(2,2,2-trifluoroacetamido)tetrahydro-2H-pyran-2-yl)oxy)-7-methoxy-6,11-dioxo-1,2,3,4,6,11-hexahydrotetracen-2-yl)ethyl pentanoate, Valrubicin, 2-oxo-2-((2S,4S)-2,5,12-trihydroxy-4-(((2S,4S,5S,6S)-5-hydroxy-6-methyl-4-(2,2,2-trifluoroacetamido)tetrahydro-2H-pyran-2-yl)oxy)-7-methoxy-6,11-dioxo-1,2,3,4,6,11-hexahydrotetracen-2-yl)ethyl pentanoate

A191480-10MG A191480-1MG A191480-25MG A191480-50MG A191480-5MG
76769-880EA 73.21 USD
76769-880 76769-882 76769-884 76769-886 76769-888
Valrubicin (AD-32) 98%
Valrubicin (AD-32)

Precaución: for Research Use Only

Formula: C₃₄H₃₆F₃NO₁₃
MW: 723.64 g/mol
MDL Number: MFCD01939322
CAS Number: 56124-62-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR