5-Bromo-4,6-dichloropyrimidin-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromo-4,6-dichloropyrimidine

A190943-1G A190943-250MG A190943-5G
76769-868EA 56.23 USD
76769-868 76769-870 76769-872
5-Bromo-4,6-dichloropyrimidin-2-amine 97%
5-Bromo-4,6-dichloropyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₄H₂BrCl₂N₃
MW: 242.9 g/mol
MDL Number: MFCD00275455
CAS Number: 7781-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR