Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II) 98%

Supplier: Ambeed
Warning

Synonyms: [1,2-Bis(diphenylphosphino)ethane]dichlorocobalt(II)

A188763-5G A188763-250MG A188763-25G A188763-100G A188763-1G
76769-816EA 238 USD
76769-816 77600-528 77600-530 77625-900 77877-418
Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II) 98%
Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II)

Precaución: for Research Use Only

Formula: C₂₆H₂₄Cl₂CoP₂
MDL Number: MFCD00134173
CAS Number: 18498-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR