2,5-Dimethylthiophene-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dimethyl-3-formylthiophene

A188075-10G A188075-1G A188075-5G
76769-810EA 150.65 USD
76769-810 76769-812 76769-814
2,5-Dimethylthiophene-3-carboxaldehyde 98%
2,5-Dimethylthiophene-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₈OS
MW: 140.203 g/mol
MDL Number: MFCD03426895
CAS Number: 26421-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR