Methyl 5-fluoro-1H-indole-2-carboxylate 95%

Supplier: Ambeed
Warning

A186967-10G A186967-250MG
76769-794EA 429.67 USD
76769-794 76769-796
Methyl 5-fluoro-1H-indole-2-carboxylate 95%
Methyl 5-fluoro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.174 g/mol
MDL Number: MFCD04972072
CAS Number: 167631-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR