6-Methoxy-4-chromanone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxychroman-4-one

A186956-100MG A186956-10G A186956-1G A186956-250MG A186956-25G A186956-5G
76769-782EA 18.04 USD
76769-782 76769-784 76769-786 76769-788 76769-790 76769-792
6-Methoxy-4-chromanone 98%
6-Methoxy-4-chromanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
MDL Number: MFCD00190159
CAS Number: 5802-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR