6-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)hexanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-(1,3-Dioxoisoindolin-2-yl)hexanoic acid

A186741-100MG A186741-1G A186741-250MG A186741-5G
76769-768EA 33 USD
76769-768 76769-770 76769-772 76769-774
6-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)hexanoic acid 98%
6-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)hexanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
MDL Number: MFCD00233499
CAS Number: 4443-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR