Methyl-5-bromo-2-methyl-3-nitrobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-bromo-2-methyl-3-nitrobenzoate
A185909-100G
A185909-10G
A185909-1G
A185909-25G
A185909-5G
76769-730EA
283.27
USD
76769-730
76769-732
76769-734
76769-736
76769-738
Methyl-5-bromo-2-methyl-3-nitrobenzoate 97%
Methyl-5-bromo-2-methyl-3-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrNO₄ MW: 274.08 g/mol |
MDL Number:
MFCD07781280 CAS Number: 220514-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...