6-Chloro-2-methylbenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A184722-100MG A184722-250MG A184722-1G
76769-692EA 25.47 USD
76769-692 76769-694 77600-488
6-Chloro-2-methylbenzo[d]thiazole 95%
6-Chloro-2-methylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNS
MW: 183.65 g/mol
MDL Number: MFCD00272359
CAS Number: 4146-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR