5-(2-Hydroxyethyl)pyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A184688-1G A184688-250MG
76769-688EA 54.11 USD
76769-688 76769-690
5-(2-Hydroxyethyl)pyrimidine-2,4(1H,3H)-dione 95%
5-(2-Hydroxyethyl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MDL Number: MFCD00627681
CAS Number: 23956-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR