Bis(phenylsulfonyl)methane 98%

Supplier: Ambeed
Warning

A184657-10G A184657-1G A184657-25G A184657-5G
76769-680EA 130 USD
76769-680 76769-682 76769-684 76769-686
Bis(phenylsulfonyl)methane 98%
Bis(phenylsulfonyl)methane

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₄S₂
MDL Number: MFCD00007553
CAS Number: 3406-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR