6-Amino-1-benzyl-3-methylpyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A184487-100MG A184487-250MG
76769-676EA 20.16 USD
76769-676 76769-678
6-Amino-1-benzyl-3-methylpyrimidine-2,4(1H,3H)-dione 95%
6-Amino-1-benzyl-3-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃O₂
MW: 231.25 g/mol
MDL Number: MFCD03480188
CAS Number: 53681-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR