Benzo[b]thiophen-4-ol 95%

Supplier: Ambeed
Warning

A184139-100MG A184139-1G A184139-250MG A184139-5G
76769-648EA 62.6 USD
76769-648 76769-650 76769-652 76769-654
Benzo[b]thiophen-4-ol 95%
Benzo[b]thiophen-4-ol

Precaución: for Research Use Only

Formula: C₈H₆OS
MW: 150.198 g/mol
MDL Number: MFCD01663547
CAS Number: 3610-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR