5-Methoxy-1H-benzimidazol-2-amine 97%

Supplier: Ambeed
Warning

A183052-1G A183052-5G
76769-622EA 33 USD
76769-622 76769-624
5-Methoxy-1H-benzimidazol-2-amine 97%
5-Methoxy-1H-benzimidazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃O
MDL Number: MFCD02752050
CAS Number: 6232-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR