5-Methyl-4-bromothiophen-2-ylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-5-methylthiophene-2-boronic acid, (4-Bromo-5-methylthiophen-2-yl)boronic acid

A182352-100MG A182352-1G A182352-250MG A182352-5G
76769-604EA 24 USD
76769-604 76769-606 76769-608 77598-242
5-Methyl-4-bromothiophen-2-ylboronic acid 95%
5-Methyl-4-bromothiophen-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₅H₆BBrO₂S
MDL Number: MFCD09952079
CAS Number: 154566-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR