5-(2-Thienyl)isoxazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-(Thiophen-2-yl)isoxazole-3-carboxylic acid

A182060-100MG A182060-1G A182060-250MG
76769-584EA 27.59 USD
76769-584 76769-586 76769-588
5-(2-Thienyl)isoxazole-3-carboxylic acid 95%
5-(2-Thienyl)isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₃S
MW: 195.192 g/mol
MDL Number: MFCD04113955
CAS Number: 763109-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR