2-Methoxy-6-(methylamino)pyridine 98%

Supplier: Ambeed
Warning

A181633-10G A181633-1G A181633-25G A181633-5G
76769-562EA 25 USD
76769-562 76769-564 76769-566 76769-568
2-Methoxy-6-(methylamino)pyridine 98%
2-Methoxy-6-(methylamino)pyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MDL Number: MFCD00191557
CAS Number: 88569-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR