2-Chloro-4-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A180069-100G A180069-10G A180069-25G A180069-500G
76769-532EA 94.43 USD
76769-532 76769-534 76769-536 76769-538
2-Chloro-4-fluorobenzonitrile 98%
2-Chloro-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClFN
MW: 155.56 g/mol
MDL Number: MFCD00042523
CAS Number: 60702-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR