2-Bromo-4-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A179463-100G A179463-10G A179463-1G A179463-25G A179463-5G
76769-510EA 175 USD
76769-510 76769-512 76769-514 76769-516 76769-518
2-Bromo-4-(trifluoromethyl)benzonitrile 97%
2-Bromo-4-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrF₃N
MDL Number: MFCD06659467
CAS Number: 35764-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR