4-tert-Butylbenzyl alcohol 98%

Supplier: Ambeed
Warning

A178766-10G A178766-1G A178766-25G A178766-5G
76769-494EA 11 USD
76769-494 76769-496 76769-498 76769-500
4-tert-Butylbenzyl alcohol 98%
4-tert-Butylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MDL Number: MFCD00004662
CAS Number: 877-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR